Ammoniumfluorid 98+% ACS

Lieferant: Thermo Scientific
Gefahr

Synonyme: Fluoroammonium

423290050 423295000 423290025
ACRO423290050EA 21.6 EUR
ACRO423290050 ACRO423295000 ACRO423290025
Ammoniumfluorid 98+% ACS
Ammonium fluoride
Formel: NH₄F
Molecular Weight: 37,04 g/mol
Schmelzpunkt: 160 °C
Dichte: 1,0088 to 1.0096 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011423
CAS-Nummer: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Residue after ignition ≤0.01 %
Titration excess NaOH ≥98.0 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR