Ammoniumfluorid 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Fluoroammonium

A16279.22 A16279.36
A16279.22EA 25.3 EUR
A16279.22 A16279.36
Ammoniumfluorid 96%
Ammonium fluoride

Purity: 96% CAS No.: 12125-01-8 Molecular Formula: FH4N Formula Weight: 37.04 Odour: Odourless Merck Reference: 14,522 MDL No.: MFCD00011423

Formel: NH₄F
Molecular Weight: 37,04 g/mol
Schmelzpunkt: 160 °C
Dichte: 1,01
Lagertemperatur: Raumtemperatur
MDL: MFCD00011423
CAS-Nummer: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 14,522

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR