Mineral oil light, Flüssigkeit für die Molekularbiologie

Lieferant: MP Biomedicals

ICNA0219483605EA 18.1 EUR
ICNA0219483605 ICNA0219483625 ICNA0219483690
Mineral oil light, Flüssigkeit für die Molekularbiologie
Mineral oil light

Mineral Oil is a colourless, odourless mixture of alkanes from a mineral source. It is often used as an inert overlay to minimize evaporation in molecular biology applications, including PCR, but has many other applications on other research fields.


  • Refractive Index: n20/D 1.467 (lit.)


Mineral oil is suitable for use as an overlay to control evaporation and cross contamination of samples in a variety of molecular biology applications.

Siedepunkt: 218,..643 °C (1013 hPa) (Lit.)
Dichte: 0,81…0.894 (15 °C) (Lit.)
MDL: MFCD00131611
CAS-Nummer: 8042-47-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR