5-Bromisochinolin, Sigma-Aldrich®

Lieferant: Merck
Achtung

594261-1G 594261-5G
SIAL594261-1GEA 26 EUR
SIAL594261-1G SIAL594261-5G
5-Bromisochinolin, Sigma-Aldrich®
5-Bromoisoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD01646405
CAS-Nummer: 34784-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR