3-Methylisochinolin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
SIAL129895-1GEA
68.2
EUR
SIAL129895-1G
3-Methylisochinolin, Sigma-Aldrich®
3-Methylisoquinoline
Formel:
C₁₀H₉N Molecular Weight: 143,19 g/mol Siedepunkt: 251 °C (1013 hPa) Schmelzpunkt: 66…67 °C |
MDL:
MFCD00024139 CAS-Nummer: 1125-80-0 EINECS: 214-412-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...