Praseodymtrifluorid

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT006553-0050EA 0 EUR
BIOT006553-0050
Praseodymtrifluorid
Praseodymtrifluorid
Formel: F₃Pr
Molecular Weight: 197,9 g/mol
Schmelzpunkt: 1395 °C
Dichte: 6,3 g/cm³ (20 °C)
MDL: MFCD00016287
CAS-Nummer: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR