Octachlornaphthalin analytischer Standard, Supelco®

Lieferant: Merck
Achtung

442725.EA 78.3 EUR
442725.
Octachlornaphthalin analytischer Standard, Supelco®
Octachlornaphthalin
Formel: C₁₀Cl₈
Molecular Weight: 403,73 g/mol
CAS-Nummer: 2234-13-1
EINECS: 218-778-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR