Pentacen, Sigma-Aldrich®
Lieferant: Merck
SIAL76440-1GEA
1090
EUR
SIAL76440-1G
SIAL76440-250MG
Pentacen, Sigma-Aldrich®
Pentacen
Formel:
C₂₂H₁₄ Molecular Weight: 278,35 g/mol Schmelzpunkt: 300 °C Dichte: 1,3 g/cm³ (20 °C) |
MDL:
MFCD00003710 CAS-Nummer: 135-48-8 EINECS: 205-193-7 Merck Index: 12,07240 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...