9-Methyladenin

Lieferant: CHEMPUR

Synonyme: 6-Amino-9-methylpurin

CHMPFL10214.POREA 0 EUR
CHMPFL10214.POR
9-Methyladenin
9-Methyladenin
Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
Schmelzpunkt: 302…304 °C
Dichte: 1,508 g/cm³ (20 °C)
MDL: MFCD00047232
CAS-Nummer: 700-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR