9-Methyladenin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Amino-9-methylpurin

M-Clarity™ quality level = MQ100

SIAL547328-250MGEA 91.5 EUR
SIAL547328-250MG
9-Methyladenin, Sigma-Aldrich®
9-Methyladenin
Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
Schmelzpunkt: 302…304 °C
Dichte: 1,508 g/cm³ (20 °C)
MDL: MFCD00047232
CAS-Nummer: 700-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR