2-Amino-3-brom-5-methylpyridin

Lieferant: MENAI (PART OF CHEMPUR)
Achtung

Synonyme: 3-Brom-5-methyl-2-pyridylamin2-Amino-3-brom-5-picolin3-Brom-5-methylpyridin-2-amin

MENALEH1.100EA 0 EUR
MENALEH1.100
2-Amino-3-brom-5-methylpyridin
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR