2-Amino-3-brom-5-methylpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-Brom-5-methyl-2-pyridylamin2-Amino-3-brom-5-picolin3-Brom-5-methylpyridin-2-amin

M-Clarity™ quality level = MQ100

SIAL543071-1GEA 32.7 EUR
SIAL543071-1G SIAL543071-5G
2-Amino-3-brom-5-methylpyridin, Sigma-Aldrich®
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR