2-Bromadamantan, Sigma-Aldrich®
Lieferant: Merck
SIAL153834-25GEA
142
EUR
SIAL153834-25G
SIAL153834-5G
2-Bromadamantan, Sigma-Aldrich®
2-Bromadamantan
Formel:
C₁₀H₁₅Br Molecular Weight: 215,13 g/mol Siedepunkt: 230 °C (1013 hPa) Schmelzpunkt: 138…140 °C |
MDL:
MFCD00074745 CAS-Nummer: 7314-85-4 EINECS: 230-777-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...