Salpetersäure 65%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Collection
Gefahr

VWRS20429.321EA 0 EUR
VWRS20429.321
Salpetersäure 65%, AnalaR NORMAPUR® analytisches Reagens
Salpetersäure
Formel: HNO₃
Molecular Weight: 63,01 g/mol
Siedepunkt: 120,5 °C (1013 hPa)
Schmelzpunkt: –42 °C
Dichte: 1,4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Bestellen

Spezifikation Testergebnisse

Assay Min. 65.0 %
Heavy metals (as Pb) Max. 0.2 ppm
Ignition residue (SO4) Max. 5 ppm
Silica Max. 0.2 ppm
Cl (Chloride) Max. 0.5 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 1 ppm
Al (Aluminium) Max. 0.1 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.05 ppm
Cu (Copper) Max. 0.05 ppm
Fe (Iron) Max. 0.2 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.2 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 3 ppm
Ni (Nickel) Max. 0.05 ppm
Pb (Lead) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR