Agar, ultrarein für die Bakteriologie
Lieferant: Thermo Scientific
Synonyme:
Agar Agar
J10906.Q1
J10906.P5
J10906.Q1EA
1697
EUR
J10906.Q1
J10906.P5
Agar, ultrarein für die Bakteriologie
Agar
Agar is used extensively as a gelling agent in the preparation of bacteriological culture media.
Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.
Schmelzpunkt:
90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081288 CAS-Nummer: 9002-18-0 EINECS: 232-658-1 Merck Index: 14,00184 |
Spezifikation Testergebnisse
Assay Gel Point (1.5%) | 32 - 38 °C |
Assay Gel Strength (1.5%) | 550 - 950 gm/cm² |
Assay Gel pH (1.5%) after Autoclaving | 6.0 - 7.5 |
Ash | ≤6.5% |
Residual Water | ≤12% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...