Vitamin B12 (Cyanocobalamin) 96,0-102,0% (durch UV-Methode) Ph. Eur., pharmazeutischer Qualität
Lieferant: PanReac AppliChem
A8454.0001EA
155
EUR
A8454.0001
Vitamin B12 (Cyanocobalamin) 96,0-102,0% (durch UV-Methode) Ph. Eur., pharmazeutischer Qualität
Vitamin B12 (Cyanocobalamin)
Formel:
C₆₃H₈₈CoN₁₄O₁₄P Molecular Weight: 1355,38 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00151092 CAS-Nummer: 68-19-9 EINECS: 200-680-0 |
Spezifikation Testergebnisse
Assay (UV, Calculated on Dried Subsance) | 96.0 - 102.0% |
Identity | Passes Test |
Loss on Drying (2 h; 105 °C; Vacuum) | Max. 12% |
Related Substance (HPLC) | Passes Test |
Absorption Maxima (278, 361, 547 - 559 nm) | Passes Test |
A (1 cm/00025%, H₂O) 361/278 nm | 1.70 - 1.90 |
A (1 cm/00025%, H₂O) 361/547 - 559 nm | 3.15 - 3.45 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...