The complete Medezine 5000 system is capable of handling health endangering dusts and protecting the user from harmful debris.
Redesigned power supply to give more power to the hand piece to cope with increased demand
Top mounted power lead for easy removal of the hand piece
New hand piece storage position to improve stability
Larger chassis wheels for easier manoeuvrability
The hand piece now has heavy duty bearings to give a longer life
The hand piece is hermetically sealed and is watertight to IP67 allowing easy washing and disinfection so the enclosed motor cannot become contaminated. All external materials used in its construction are non corrosive.