(-)-Epigallocatechin gallate ≥98%, kristalliner Feststoff
Lieferant: Cayman Chemical
Synonyme:
(-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG
70935-100
70935-50
70935-500
70935-250
CAYM70935-100EA
91
EUR
CAYM70935-100
CAYM70935-50
CAYM70935-500
CAYM70935-250
(-)-Epigallocatechin gallate ≥98%, kristalliner Feststoff
(-)-Epigallocatechin gallate
(–)-Epigallocatechin gallate (EGCG) is a phenol, found in a variety of plants including green and black tea plants and has diverse biological activities.
Formel:
C₂₂H₁₈O₁₁ Molecular Weight: 458,38 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00075940 CAS-Nummer: 989-51-5 |
Spezifikation Testergebnisse
Purity | ≥98% |
λmax | 276 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...