ELISA Kit, euroProxima lactoferrin, r-biopharm®
Lieferant: R-Biopharm
RBIO5091LFEREA 1160 EUR
RBIO5091LFER
ELISA Kit, euroProxima lactoferrin, r-biopharm®
Tests ELISAs
A competitive enzyme immunoassay for screening and quantitative analysis of lactoferrin in various matrices.
  • Test format: Microtiter plate, 96 wells
  • Detection limit: UHT 5 μg/ml; baby and infant milk powder 125 μg/ml
The Lactoferrin ELISA is used for the quantification of the bioactive bovine milk protein (80 kDa) in milk and milk products. Lactoferrin plays a key role in the defense mechanisms of lactating animals’ mammary glands.