Biocytin TMR, CF® 568 Dye
Lieferant: Biotium
CF®568, an Outshines Alexa Fluor®568
- Yields much brighter antibody conjugates than Alexa Fluor® 568
- Extremely photostable
- Excellent choice for multi-color imaging with CF®488A and CF®640R
- Compatible with TIRF and multicolor STORM superresolution imaging
CF® dye biocytin is a cell-impermeant fixable polar tracer that is a conjugate of CF® dye and biotin with an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis.
Stock solutions may be prepared in DMF or DMSO.
CF® dye biocytin is available in the following colors: CF®488A, CF®568, CF®594, and CF®640R.
Bitte beachten Store at –20 °C, protected from light.
Spezifikation Testergebnisse
Abs/Em Maxima | 562/583 nm |
Extinction coefficient | 100,000 |
Molecular weight | ~ 714 |
Excitation laser line | 532 nm or 568 nm |
Direct replacement for | Alexa Fluor® 568, ATTO 565, Rhodamine Red |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...