Hämatoxylin-Lösung (Mayer), Clin-Tech

Lieferant: KLINIPATH (PL)
Achtung

Synonyme: HydroxybrasilinNaturschwarz 1

VWRK640400EA 137 EUR
VWRK640400
Hämatoxylin-Lösung (Mayer), Clin-Tech
Hämatoxylin-Färbelösung

Haemalum (Mayer’s)


Contents: Haematoxylin 0.1%, with Citric acid / Potassium Aluminium Sulphate / Chloral hydrate


Bancroft Theory & Practice of Histological Techniques 6th Ed. p123

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
MDL: MFCD00078111
CAS-Nummer: 517-28-2
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR