AEROSIL® R 972

Lieferant: OVERLACK CHEMISCHE PRODUKTE

OVERAEROSILR972EA 512 EUR
OVERAEROSILR972
AEROSIL® R 972
AEROSIL®
MDL: MFCD00011232
CAS-Nummer: 112945-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR