Organic reference standard, 2,5-Dihydroxybenzoic acid, analytical standard, Supelco®
Lieferant: Merck
78069-25MGEA 51.9 EUR
78069-25MG
Organic reference standard, 2,5-Dihydroxybenzoic acid, analytical standard, Supelco®
Standards Organische Standards
2,5-Dihydroxybenzoic acid may be used as a reference standard in the determination of 2,5-dihydroxybenzoic acid in biopsy samples using high-performance liquid chromatography coupled with electrochemical detector.