Organic reference standard, Ethylhexyl triazone, analytical standard, Supelco®
Lieferant: Merck
93465-100MGEA 91.7 EUR
93465-100MG
Organic reference standard, Ethylhexyl triazone, analytical standard, Supelco®
Standards Organische Standards
Ethylhexyl triazone is a ultraviolet-B filter, which is used as a sunscreen agent in skin care products.
Verpackung: Bottomless glass bottles are inserted with fused cone.