HMDS (1,1,1,3,3,3-Hexamethyldisilazan) Silylierungsreagenz, zur GC-Derivatisierung
Lieferant: MACHEREY-NAGEL
Gefahr
Synonyme: Bis(trimethylsilyl)amin
MANA701240.510EA 55.3 EUR
MANA701240.510 MANA701240.650
HMDS (1,1,1,3,3,3-Hexamethyldisilazan) Silylierungsreagenz, zur GC-Derivatisierung
HMDS (1,1,1,3,3,3-Hexamethyldisilazan)
HMDS is weak TMS donor. If used as sole reagent, it is slow and not very effective. After addition of catalytic quantities (e.g., 1 %) of TMCS or as a mixture with TMCS it is a fast and quantitative reagent for trimethylsilylation of organic compounds.
  • Silylation can be applied on many compounds
  • Silylation reagents are easily prepared
  • HMDS reagent mixed with TMCS catalyst shows fast and quantitative reaction
Aprotic solvents like acetonitrile, pyridine, dimethylformamide, carbon disulfide and dimethylacetamide are recommended for use with HMDS.
Formel: (CH₃)₃SiNHSi(CH₃)₃
Molecular Weight: 161,39 g/mol
Siedepunkt: 126 °C (760 mm Hg)
Dichte: 0,7742 g/cm³ (d20°/4°)
MDL: MFCD00008259
CAS-Nummer: 999-97-3
UN: 3316
ADR: 9,II
https://de.vwr.com/stibo/web/std.lang.all/80/76/2298076.jpg