Organic reference standard, Benzyl benzoate, analytical standard, Supelco®
Lieferant: Merck
68183-1MLEA 62.6 EUR
68183-1ML
Organic reference standard, Benzyl benzoate, analytical standard, Supelco®
Standards Organische Standards
Benzyl benzoate undergoes hydrogenation reaction catalyzed by a system of (Ru(acac)₃) (Where Ru-ruthenium/acac-acetylacetonate) with MeC(CH₂PPh₂)₃ (tripodal triphosphine ligand).
In the presence of excess of triethylamine in trifluoroethanol obtain benzyl alcohol.