Dietary Fiber, total, assay control kit, Supelco®

Lieferant: Merck
TDFC10-1KTEA 590 EUR
TDFC10-1KT
Dietary Fiber, total, assay control kit, Supelco®
Standards Lebensmittel-Standards
Standards for use as internal controls in conjunction with the Total Dietary Fiber Assay Kit (TDF100A).

Arabinogalactan (stractan) 1 g
Casein 5 g
β-Glucan 1 g
Pectin 1 g
Starch, corn 10 g
Starch, wheat 10 g
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR