Sulphorhodamine 101 fluoreszierender Farbstoff, für die Histologie

Lieferant: Biotium

80101.EA 325 EUR
80101.
Sulphorhodamine 101 fluoreszierender Farbstoff, für die Histologie
Sulphorhodamine 101

Sulforhodamine 101 has a longer absorption wavelength than does sulforhodamine B. Like sulforhodamine B and G, in addition to their potential use in cancer drug screening, these fluorescent dyes have been primarily used as polar tracers for the studies of neuronal cell morphology and cell-cell communication.


  • λEx/λEm = 586/605 nm
  • Red solid soluble in water
  • Store at room temperature

Formel: C₃₁H₃₀N₂O₇S₂
Molecular Weight: 606,708 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012407
CAS-Nummer: 60311-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR