Biotin ethylenediamine hydrobromide
Lieferant: Biotium
Synonyme:
Neurobiotin™
90057.EA
163
EUR
90057.
Biotin ethylenediamine hydrobromide
Biotin ethylenediamine hydrobromide
Biotin ethylenediamine, hydrobromide (N-(5-(6-((biotinoyl)amino)hexanoyl)amino)pentylmaleimide) is identical to Neurobiotin™, a useful anterograde and transneuronal tracer.
Neurobiotin is a trademark of Vector Laboratories.
Formel:
C₁₂H₂₃BrN₄O₂S Molecular Weight: 367,31 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00467807 CAS-Nummer: 216299-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...