Sigma Pseudo™ Narcotic Scent Heroin formulation, powder, Supelco®
Lieferant: Merck
P2548-100G
P2548-100GEA 58.8 EUR
P2548-100G
Sigma Pseudo™ Narcotic Scent Heroin formulation, powder, Supelco®
Standards Biologische Standards
Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of heroin.
Bitte beachten Storage temperature: −20 °C