Amberlite® IRA-900 (Cl), ca. 60% Feuchtigkeit Ionenaustauscherharz, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

216585-500GEA 131 EUR
216585-500G
Amberlite® IRA-900 (Cl), ca. 60% Feuchtigkeit Ionenaustauscherharz, Supelco®
Amberlite® IRA-900 (Cl)

Amberlite® IRA-900 chloride form is a macroporous, polystyrenic type 1 resin.[1] This resin was found to catalyze the photodecomposition of carbon tetrachloride in ethanol.


Strongly basic, macroreticular resin of moderately high porosity with benzyltrialkylammonium functionality which is useful in decolorizing and removal of organic materials.


Amberlite® IRA-900 modified with metallophthalocyanines was used to remove 4-nitrophenol.


Strongly basic (type I) anion exchange resin for decolorizing intermediate to light solutions, concentrating heparin at neutral pH, removing metal ions, etc.

MDL: MFCD00132712
CAS-Nummer: 9050-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR