Trichrome stain Kit (Masson)
Lieferant: RAL (REACTIFS RAL SA)
Staining for Histological and Cytological studies of Fixed and Paraffined Tissue Sections.
Staining with kit Trichrome de Masson, Blue aniline variation enables a trichromic station on histological sections with a specific coloration of the collagen by Blue.
Mayer Haemalum stains nuclei black-blue or brown. Ponceau Fuchsin solution stains cytoplams red pink. Light green stains collagen blue.
Components - Name (ref) Packing
MAYER HAEMALUM (Ref 320550) 100 ml (Others packagings 1000, 2500 ml)
FUCHSIN PONCEAU (Ref 361770) 100 ml (Others packagings 1000, 2500 ml)
1% ACETIC WATER (Ref 361840) 200 ml
ACID PHOSPHOMOLYBDIQUE (Ref 300140) 100 ml
ANILINE BLUE (Ref 361880) 100 ml
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...