3-Amino-N-methylbenzamid ≥98%

Lieferant: ABCR GMBH
Achtung

ABCRTCA1067.0025EA 0 EUR
ABCRTCA1067.0025
3-Amino-N-methylbenzamid ≥98%
3-Amino-N-methylbenzamid
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
Schmelzpunkt: 119°C
MDL: MFCD00070630
CAS-Nummer: 25900-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR