3-Ethoxypropionsäure

Lieferant: ABCR GMBH

ABCRTCE0053.0025EA 0 EUR
ABCRTCE0053.0025
3-Ethoxypropionsäure
3-Ethoxypropionsäure
Formel: C₅H₁₀O₃
MDL: MFCD00058997
CAS-Nummer: 4324-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR