Cell culture supplements, Panexin NTA serum substitute
Lieferant: PAN-Biotech
PANBP04-95750EA 278.8 EUR
PANBP04-95070 PANBP04-95090 PANBP04-95700P PANBP04-95750 PANBP04-95070P
Cell culture supplements, Panexin NTA serum substitute
Zellkultur-Ergänzungen und -Zusätze
Panexin NTA is a ready-to-use, fully defined serum substitute for the cultivation of adherent cells under serum-free conditions. This solution is added to the culture medium in a final concentration of 10%. It supports the adherent growth of many cell types in an optimum manner.
  • Sterile solution
  • Store at ‒20 °C
Panexin NTA contains purified proteins, lipids, salts, amino acids, trace elements, attachment factors and hormones in an optimised formulation. Panexin NTA contains no growth factors, undefined hydrolysates or peptones.
Panexin NTA Pharma grade is free of any animal-derived components.
It has been shown for many cell lines that Panexin NTA can fully replace FBS. Due to specially selected and pretested raw materials Panexin NTA batches are very homogeneous, therefore the complex batch testing known from FBS can be omitted with the use of Panexin NTA.
Panexin NTA is fully defined and contains no undefined peptones or hydrolysates. Due to this special formulation the interpretation of results from studies on effects of individually added growth factors is easier and more reliable in serum-free conditions. For cell lines which require specific growth factors these should be added in a concentration as previously used.