Bovine serum albumin (BSA)
Lieferant: PAN-Biotech
PANBP06-138850EA 320.1 EUR
PANBP06-138850
Bovine serum albumin (BSA)
Proteine und Peptide
Albumins serve as additive proteins for tissue cultures. They are the main protein component in serum and are added to cell culture media to increase the stability of cell membranes and to bind possibly toxic elements.
  • Minimal lot to lot variation
  • Stringent control of raw materials
  • Minimal BSE/TSE risk
  • US origin
  • Full documentation
  • Special products for unique applications