Cell culture media, Dulbecco's modified eagle medium (DMEM)
Lieferant: PAN-Biotech
PANBP04-01507EA 37.3 EUR
PANBP03-0051 PANBP03-1001 PANBP03-0110 PANBP03-0450 PANBP04-03601 PANBP04-03588S PANBP04-01546S1 PANBP04-01546S2 PANBP04-01507 PANBP04-01159S1 PANBP04-03500S3 PANBP03-0801 PANBP04-01504 PANBP04-01603 PANBP04-01348 PANBP04-02513 PANBP04-03160 PANBP04-035501 PANBP04-01166 PANBP04-04054 PANBP04-05553 PANBP04-01250 PANBP04-015010 PANBP04-03510 PANBP03-00150 PANBP04-04502 PANBP04-03612 PANBP04-04507 PANBP04-04508 PANBP04-01550S3 PANBP04-01550S1 PANBP04-01557 PANBP04-01512 PANBP04-01556 PANBP04-01511 PANBP04-03596S2 PANBP03-0210 PANBP04-03593 PANBP03-01501
Cell culture media, Dulbecco's modified eagle medium (DMEM)
Zellkulturmedien
Dulbecco's Modified Eagle Medium (DMEM) is a modification of Basal Medium Eagle (BME) which is Intrinsically developed for the cultivation of murine embryonic cells. DMEM is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS.
  • Suitable for most cell types
  • Available in a broad variety of formulations
  • Also available with stable glutamine (L-Alanyl-L-Glutamine) for a better shelf life
These media contain approximately four times as much of the vitamins and amino acids, and two to four times as much glucose depending on the version as the original formulation.
DMEM with 1,0 g/l Glucose is the standard medium, whereas DMEM with 4,5 g/l Glucose is for cells which have a high energy demand.