Filtrated bovine serum, FBS Good, EU approved
Lieferant: PAN-Biotech
PANBP40-37500EA 160 EUR
PANBP40-37500 PANBP40-37100
Filtrated bovine serum, FBS Good, EU approved
Seren
FBS Good is a naturally defined serum with a sustained growth promoting property and a higher safety.
  • Reproducible growth properties
  • Very low endotoxin level
  • Suitable for a great variety of cells
  • Continuous quality control
  • 0,2 µm sterile filtered
  • Store at ‒20 °C
FBS Good only contains serum of highest quality from defined countries as specified. It is not blended or enhanced by addition of growth factors or proteins.