Cell culture media, Dulbecco's modified eagle medium/F12 Ham's (DMEM:F12)
Lieferant: PAN-Biotech
PANBP04-41250EA 15.3 EUR
PANBP03-1250 PANBP03-1150 PANBP03-1210 PANBP03-6110 PANBP03-6010 PANBP03-1110 PANBP03-6420 PANBP04-41506 PANBP03-6150 PANBP03-6050 PANBP04-41552 PANBP04-41662 PANBP04-41661 PANBP04-41154 PANBP04-41231 PANBP04-41250 PANBP04-41505 PANBP04-41504 PANBP04-41658 PANBP03-1101 PANBP04-41258 PANBP04-41752
Cell culture media, Dulbecco's modified eagle medium/F12 Ham's (DMEM:F12)
Zellkulturmedien
This media combines the advantages of both media DMEM (high concentration of amino acids and vitamins) and Ham’s F12 (higher concentration of zinc sulfate, putrescine and linoleic acid).
  • Suitable for clonal density cultures
  • This medium supports the growth of almost all cell lines
These are used for pancreas cells, Sertoli cells or to culture cells, which are used for human protein production.