G418 Disulfat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat

M-Clarity™ quality level = MQ200

G1279-1G G1279-250MG G1279-5G
SIALG1279-1GEA 661 EUR
SIALG1279-1G SIALG1279-250MG SIALG1279-5G
G418 Disulfat, Sigma-Aldrich®
G418 disulphate
Formel: C₂₀H₄₄N₄O₁₈S₂
Molecular Weight: 692,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058314
CAS-Nummer: 108321-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR