Cell culture media, Waymouth's MB 752/1
Lieferant: PAN-Biotech
PANBP04-28500EA 95.5 EUR
PANBP03-4510 PANBP03-4550 PANBP04-28500
Cell culture media, Waymouth's MB 752/1
Zellkulturmedien
Waymouth's is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment.
  • Used for growing strain L sublines, NCTC clone 929
The Waymouth's MB 752/1 media was developed for studies concerning nutrition and metabolism.