(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
(S)-(-)-BINAP
STRE15-0151-5GEA
695
EUR
STRE15-0151-5G
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
BINAP, Phosphine
Formel:
C₄₄H₃₂P₂ Molecular Weight: 622,69 g/mol Schmelzpunkt: 241…242 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010805 CAS-Nummer: 76189-56-5 Merck Index: 13,01223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...