Methyltridecanoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tridecansäuremethylester
M-Clarity™ quality level = MQ200
SIALT0627-1GEA
67.9
EUR
SIALT0627-1G
PFIZT-0627
SIALT0627-10G
Methyltridecanoat, Sigma-Aldrich®
Methyltridecanoat
Formel:
C₁₄H₂₈O₂ Molecular Weight: 228,38 g/mol Siedepunkt: 290 °C (1013 hPa) Schmelzpunkt: 5,5 °C Dichte: 0,864 g/cm³ (25 °C) |
MDL:
MFCD00008977 CAS-Nummer: 1731-88-0 EINECS: 217-054-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...