2-[Bis(2-hydroxyethyl)amino]ethansulfonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
BES, N,N-Bis-(2-hydroxyethyl)-2-aminoethansulfonsäure
M-Clarity™ quality level = MQ400
B4554-100G
B4554-1KG
B4554-25G
B4554-500G
SIALB4554-100GEA
307
EUR
SIALB4554-100G
SIALB4554-1KG
SIALB4554-25G
SIALB4554-500G
2-[Bis(2-hydroxyethyl)amino]ethansulfonsäure, Sigma-Aldrich®
2-[Bis(2-hydroxyethyl)amino]ethansulfonsäure
Formel:
C₆H₁₅NO₅S Molecular Weight: 213,25 g/mol Siedepunkt: 405 °C (1013 hPa) Schmelzpunkt: 152…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007533 CAS-Nummer: 10191-18-1 EINECS: 233-465-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...