MOPS-Na, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Natrium-4-morpholin-1-ylpropylsulfonat3-(N-Morpholino)propansulfonsäure NatriumsalzMOPS Natriumsalz

475899-100EA 149 EUR
475899-100
MOPS-Na, Millipore®
MOPS-Na
Formel: C₇H₁₄NNaO₄S
Molecular Weight: 231,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064350
CAS-Nummer: 71119-22-7
EINECS: 428-420-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR