4-(Dihexylamino)benzoesäure

Lieferant: ABCR GMBH

ABCRAB145157.0001EA 0 EUR
ABCRAB145157.0001
4-(Dihexylamino)benzoesäure
4-(Dihexylamino)benzoesäure
Formel: C₁₉H₃₁NO₂
Molecular Weight: 305,46 g/mol
CAS-Nummer: 134690-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR