PIPES
Lieferant: PanReac AppliChem
Synonyme:
2,2'-(Piperazin-1,4-diyl)bis(ethansulfonsäure)
, Piperazin-1,4-bis(2-ethansulfonsäure)
, Piperazin-N,N'-bis-2-ethansulfonsäure
A1079.0500EA
300
EUR
A1079.0500
PIPES
PIPES
Formel:
C₈H₁₈N₂O₆S₂ Molecular Weight: 302,37 g/mol Siedepunkt: 499 °C (1013 hPa) Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006159 CAS-Nummer: 5625-37-6 EINECS: 227-057-6 Merck Index: 12,07633 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...