PIPES ≥99,0%, ultrareines Bioreagenz für die Molekularbiologie, J.T.Baker®
Lieferant: Avantor
Synonyme:
2,2'-(Piperazin-1,4-diyl)bis(ethansulfonsäure)
, Piperazin-1,4-bis(2-ethansulfonsäure)
, Piperazin-N,N'-bis-2-ethansulfonsäure
4265-01EA
128.96
EUR
4265-01
4265-04
PIPES ≥99,0%, ultrareines Bioreagenz für die Molekularbiologie, J.T.Baker®
PIPES
Formel:
C₈H₁₈N₂O₆S₂ Molecular Weight: 302,37 g/mol Siedepunkt: 499 °C (1013 hPa) Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006159 CAS-Nummer: 5625-37-6 EINECS: 227-057-6 Merck Index: 12,07633 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | |
Assay (C₈H₁₈N₂O₆S₂) | ≥99.0 % |
Appearance | Passes Test |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Protease Activity | Passes Test |
ACS - Heavy Metals (as Pb) | ≤5 ppm |
Insoluble Matter | ≤0.005 % |
Residue after Ignition | ≤0.30 % |
Product Information (not specifications): pKa at 20°C = 6.80 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...