Natriumperiodat, Pierce™

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Natriumtetroxoiodat(VII)Natriummetaperiodat

PIER20504EA 49.87 EUR
PIER20504
Natriumperiodat, Pierce™
Natriumperiodat

Thermo Scientific Pierce Sodium meta-Periodate is a gentle oxidizing agent that cleaves cis-diols in carbohydrate sugars to create amine-reactive aldehydes, providing many uses relating to the study and detection of glycoproteins.


  • Related applications: Protein Labeling & Crosslinking

Formel: NaIO₄
Molecular Weight: 213,89 g/mol
Siedepunkt: 300 °C (1013 hPa)
Schmelzpunkt: 175 °C
Dichte: 3,87 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003534
CAS-Nummer: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 13,08714

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR