Calciumiodid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL439797-5GEA
141
EUR
SIAL439797-5G
Calciumiodid, Sigma-Aldrich®
Calcium iodide
Formel:
CaI₂ Molecular Weight: 293,89 g/mol Siedepunkt: 1100 °C (1013 hPa) Schmelzpunkt: 740 °C Dichte: 3,956 g/cm³ (25 °C) |
MDL:
MFCD00010910 CAS-Nummer: 10102-68-8 EINECS: 233-276-8 Merck Index: 12,01720 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...